789.46 GBP valid until 2025-12-31
Use promo code "24380" to get your promotional price.
Use promo code "24380" to get your promotional price.
Product Type | Chemical Protective Clothing |
---|---|
Size | 2X-Large |
Gender | Unisex |
Material | Multi-layer Barrier Laminate Fabric |
Color | Orange |
Disposable | Reusable |
Waist Style | Elastic |
Ankle Style | Elastic,Boot Overflaps |
Ankle Style | Elastic,Boot Overflaps |
Cuff Style | Double,Elastic |
Cuff Style | Double,Elastic |
Neck Style | Collared |
Hood Type | Standard |
Closure Type | Double Zipper |
Seam Type | Welded,Taped |
Seam Type | Welded,Taped |
Sterility | Non-sterile |
Protection Class | CE CATEGORY III |
Resistance | Chemicals,Anti-static,Particle |
Resistance | Chemicals,Anti-static,Particle |
Resistance | Chemicals,Anti-static,Particle |
Certifications/Compliance (Europe) | CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5) |
No. of Pockets | None |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Unexpected leakages spills or other releases |