Use promo code "24157" to get your promotional price.
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays
Package biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe in these durable, break-resistant media bottles.
473.62 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Use promo code "24157" to get your promotional price.
Code:
55
Additional Details:
Weight:
5.42000kg
Specification
| Capacity (Metric) | 125 mL |
|---|---|
| Capacity (English) | 4.2 oz. |
| Color | Transparent |
| Description | With septum closure |
| Diameter (English) | 2.13 in. |
| Diameter (English) Inner Neck | 1.1 in. |
| Diameter (Metric) Inner Neck | 28 mm |
| Material | PETG |
| No. per Case | 96 |
| No. per Pack | 24 |
| Sterility | Sterile |
| Certifications/Compliance | Meet USP Class IV standards. Noncytotoxic. |
| Closure Included | Yes |
| Graduated | Yes |
| Mouth | Wide |
| Shape | Square |
| Fit for Purpose | Sterile |
| Height (Metric) | 110 mm |
| Product Type | Media Bottle |
| Height (English) | 4.33 in. |
| Closure Size | 38-430 mm |
| Diameter (Metric) | 54 mm |