Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 2.5 L |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
CAS | 108-87-2 |
---|---|
Molecular Formula | C7H14 |
Molecular Weight (g/mol) | 98.19 |
MDL Number | MFCD00001497 |
InChI Key | UAEPNZWRGJTJPN-UHFFFAOYSA-N |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
PubChem CID | 7962 |
IUPAC Name | methylcyclohexane |
SMILES | CC1CCCCC1 |
Specification
Chemical Name or Material | Methylcyclohexane |
---|---|
Name Note | 99% |
CAS | 108-87-2 |
Melting Point | -126.3°C |
Density | 0.7700g/mL |
Boiling Point | 101.0°C |
Flash Point | −3°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Molecular Formula | C7H14 |
Linear Formula | C6H11CH3 |
Refractive Index | 1.4210 to 1.4230 |
MDL Number | MFCD00001497 |
Quantity | 2.5 L |
Beilstein | 05, 29 |
Merck Index | 15, 6118 |
Specific Gravity | 0.77 |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Synonym | cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan |
Solubility Information | Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol |
InChI Key | UAEPNZWRGJTJPN-UHFFFAOYSA-N |
SMILES | CC1CCCCC1 |
IUPAC Name | methylcyclohexane |
Molecular Weight (g/mol) | 98.19 |
PubChem CID | 7962 |
Formula Weight | 98.19 |
Percent Purity | 99% |
Grade | Extra Pure |
Physical Form | Liquid |
Technical documents
[Methylcyclohexane]
[Danger]
[{code=AQUA CHRONIC 2, sequence=1, description=LONG-TERM AQUATIC HAZARD Chronic 2}, {code=ASP. TOX. 1, sequence=2, description=Aspiration toxicity Category 1}, {code=FLAM. LIQ. 2, sequence=3, description=Flammable liquid Category 2}, {code=SKIN IRRIT. 2, sequence=4, description=Skin corrosion/irritation Category 2}, {code=STOT SE 3, sequence=5, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H304, sequence=2, description=May be fatal if swallowed and enters airways.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H336, sequence=4, description=May cause drowsiness or dizziness.}, {code=H411, sequence=5, description=Toxic to aquatic life with long lasting effects.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P240, sequence=52, description=Ground/bond container and receiving equipment.}, {code=P261, sequence=53, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P273, sequence=54, description=Avoid release to the environment.}, {code=P301+P310, sequence=55, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}, {code=P302+P352, sequence=56, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P331, sequence=57, description=Do NOT induce vomiting.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Methylcyclohexane}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}, {code=GHS09, sequence=4, description=ENVIRONMENT}]
Product Identifier
- Methylcyclohexane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H304- May be fatal if swallowed and enters airways.
- H315- Causes skin irritation.
- H336- May cause drowsiness or dizziness.
- H411- Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240- Ground/bond container and receiving equipment.
- P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273- Avoid release to the environment.
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P331- Do NOT induce vomiting.
Supplemental Information
- MIXTURE LIST- Contains : Methylcyclohexane