Use promo code "25339" to get your promotional price.
Potassium hydroxide, ca. 85%, for analysis, pellets
Use promo code "25339" to get your promotional price.
| Quantity | 1 kg |
|---|---|
| Packaging | Plastic bottle |
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| SMILES | [OH-].[K+] |
Specification
| Chemical Name or Material | Potassium hydroxide |
|---|---|
| Name Note | ca. 85 % |
| Boiling Point | 1320.0°C |
| CAS | 1310-58-3 |
| Melting Point | 360.0°C |
| Assay Percent Range | ca. 85% |
| Molecular Formula | HKO |
| Linear Formula | KOH |
| MDL Number | MFCD00003553 |
| Fieser | 01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258 |
| Merck Index | 15, 7761 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Synonym | potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693 |
| Solubility Information | Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C), soluble in ethanol (400g/L), soluble in ammonia, slightly soluble in ether |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Quantity | 1 kg |
| SMILES | [OH-].[K+] |
| IUPAC Name | potassium hydroxide |
| Molecular Weight (g/mol) | 56.11 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| Formula Weight | 56.11 |
| Percent Purity | ∼85% |
| Grade | Analytical |
| Packaging | Plastic bottle |
| Physical Form | Pellet |
Technical documents
[Potassium hydroxide]
[Danger]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=EYE DAM. 1, sequence=2, description=Serious eye damage/eye irritation Category 1}, {code=MET. CORR. 1, sequence=3, description=Substances and mixtures corrosive to metals Category 1}, {code=SKIN CORR. 1A, sequence=4, description=Skin corrosion/irritation Category 1 A}]
[{code=H318, sequence=1, description=Causes serious eye damage.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H302, sequence=3, description=Harmful if swallowed.}, {code=H290, sequence=4, description=May be corrosive to metals.}]
[{code=P280, sequence=50, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=51, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=52, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=54, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- Potassium hydroxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318- Causes serious eye damage.
- H314- Causes severe skin burns and eye damage.
- H302- Harmful if swallowed.
- H290- May be corrosive to metals.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.