338.18 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Use promo code "24140" to get your promotional price.
| Color | Clear |
|---|---|
| Solvent Or Alcohol Resistant | Life Science Reagent Resistant |
| Description | SEALING TAPE POLYESTR STRL CLR |
| Material | Polyester |
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| No. of Wells | 96 |
| No. per Case | 800 |
| No. per Pack | 200 |
| For Use With (Application) | Cell culture, sterile applications |
| Content And Storage | General |
| Product Type | Adhesive Seal |
| Product Line | Nunc |
| Adhesive | Acrylic |
| Certified RNase/DNase-free | No |
| Pierceable | No |
| Temperature Range (Metric) | -40°C to +120°C |
| Well Shape | All well shapes |
| Plate Compatibility | All Microplates |
| Incubation Conditions | General |
| Peelable | Yes |