184.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
| Certifications/Compliance | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
|---|---|
| Neck Style | Angled |
| No. per Case | 30 |
| No. per Pack | 5 |
| Graduated | Yes |
| Sterility | Sterile |
| Bottom Shape | Flat |
| Closure Type | Solid |
| Culture Area | 225 cm² |
| Surface Treatment | Cell Culture-treated |
| Volume (Metric) Working | 70 mL |