72.52 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Use promo code "24157" to get your promotional price.
| Capacity (English) | 1.69 oz. |
|---|---|
| Closure Material | Polyethylene |
| Capacity (Metric) | 50 mL |
| Material | Polystyrene |
| For Use With (Application) | For enhanced cell attachment and growth |
| Neck Style | Canted |
| No. per Case | 100/Cs. |
| No. per Pack | 20/Pk. |
| Shape | Rectangular |
| Surface Coating | TC-Treated |
| Sterility | Sterile |
| Bottom Shape | Rectangular |
| Closure Type | Vented |
| Culture Area | 25 cm² |