356.67 GBP valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Use promo code "24124" to get your promotional price.
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
|---|---|
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 10 Pk. |
| Product Line | Nunc |
| Color | White |
| Product Type | Microplate |
| Skirt Style | Low Flange Pinchbar |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 50 to 250 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Flat Bottom (F-well) |
| No. per Case | 180 |
| No. per Pack | 10 |