Code:
66
Falcon™ Polystyrene Microplates
Constructed of crystal grade virgin polystyrene for optimum visibility. Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.
Additional Details:
Weight:
5.32970kg
Specification
| Array | 3 x 4 |
|---|---|
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 6 mL |
| Color | Clear |
| Culture Area | 3.8 cm2 |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Type | Cell Culture Plate |
| Volume (Metric) Working | 1.5 to 2.2 mL |
| Well Count | 12 |
| Includes | Lid |
| Packaging | Individually Wrapped |
| Quantity | 50/Cs. |