31.16 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
| Quantity | 1 L |
|---|---|
| Packaging | Plastic bottle |
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
| Chemical Name or Material | Sulfuric acid |
|---|---|
| Color | Colorless |
| Name Note | 96.00% (±0.50%) Solution in Water |
| CAS | 7664-93-9 |
| Melting Point | 10.0°C |
| Boiling Point | 290.0°C |
| Density | 1.8300g/mL |
| Packaging | Plastic bottle |
| Specific Gravity | 1.83 |
| Quantity | 1 L |
| Molecular Formula | H2O4S |
| Viscosity | 21 mPa.s (25°C) |
| Linear Formula | H2SO4 |
| Physical Form | Oily Liquid |
| MDL Number | MFCD00064589 |
| Fieser | 01,470; 05,633; 06,558; 07,347; 09,441 |
| Merck Index | 15, 9104 |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Synonym | oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil |
| Solubility Information | Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat) |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| SMILES | OS(O)(=O)=O |
| IUPAC Name | sulfuric acid |
| Molecular Weight (g/mol) | 98.07 |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| Formula Weight | 98.08 |
| Percent Purity | 95.85 to 96.15% |
| Grade | Pure |
[Sulfuric acid]
[Danger]
[{code=SKIN CORR. 1A, sequence=1, description=Skin corrosion/irritation Category 1 A}, {code=EYE DAM. 1, sequence=2, description=Serious eye damage/eye irritation Category 1}]
[{code=H314, sequence=1, description=Causes severe skin burns and eye damage.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}]
[{code=GHS05, sequence=1, description=CORROSION}]