568.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
| Barcode | Custom Barcoding Available |
|---|---|
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Certified | Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility |
| Lid | With Lid |
| Array | 8 x 12 |
| Packaging | 10 Pk., 160 Cases |
| Product Line | Nunc |
| Color | Black |
| Product Type | Microplate |
| Material | Polystyrene |
| Volume (Metric) Well | 400 μL |
| No. of Wells | 96 |
| Volume (Metric) Working | 50 to 250 μL/well |
| Sterility | Sterile |
| Surface Treatment | Nunclon Delta |
| Well Shape | Flat Bottom (F-well) |
| No. per Case | 160 |
| No. per Pack | 10 |
| Quantity | 160/Cs. |