Use promo code "24128" to get your promotional price.
Sodium hypochlorite, 4-6%
Use promo code "24128" to get your promotional price.
| Quantity | 1 L |
|---|---|
| Packaging | Plastic bottle |
Chemical Identifiers
| CAS | 7681-52-9 |
|---|---|
| Molecular Formula | ClNaO |
| Molecular Weight (g/mol) | 74.44 |
| MDL Number | MFCD00011120 |
| InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| PubChem CID | 23665760 |
| ChEBI | CHEBI:32146 |
| IUPAC Name | sodium;hypochlorite |
| SMILES | [O-]Cl.[Na+] |
Specification
| Chemical Name or Material | Sodium hypochlorite |
|---|---|
| Name Note | 4-6% Chlorine |
| CAS | 7681-52-9 |
| Color | Yellow |
| CAS | 7732-18-5 |
| Physical Form | Solution |
| Quantity | 1 L |
| Molecular Formula | ClNaO |
| Linear Formula | NaOCl |
| MDL Number | MFCD00011120 |
| Fieser | 01,1084; 04,456; 05,617; 06,543; 07,337; 08,461; 09,430; 10,365; 11,487; 15,293; 16,308 |
| Merck Index | 15,8762 |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Synonym | sodium hypochlorite,antiformin,sodium oxychloride,chlorox,clorox,javex,javelle water,hypochlorous acid, sodium salt,hypochlorite sodium,carrel-dakin solution |
| Solubility Information | Solubility in water: soluble |
| InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
| SMILES | [O-]Cl.[Na+] |
| IUPAC Name | sodium;hypochlorite |
| Molecular Weight (g/mol) | 74.44 |
| PubChem CID | 23665760 |
| ChEBI | CHEBI:32146 |
| Formula Weight | 74.44 |
| Percent Purity | 4.5 to 5.5% |
| Grade | Reagent |
| Packaging | Plastic bottle |
| Specific Gravity | 1.083 |
| Density | 1.0830g/mL |
Technical documents
[Sodium hypochlorite, 5% active chlorine]
[Danger]
[{code=H290, sequence=1, description=May be corrosive to metals.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H400, sequence=3, description=Very toxic to aquatic life.}, {code=H411, sequence=4, description=Toxic to aquatic life with long lasting effects.}, {code=EUH031, sequence=91, description=Contact with acids liberates toxic gas.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Sodium hypochlorite}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS09, sequence=2, description=ENVIRONMENT}]
Product Identifier
- Sodium hypochlorite, 5% active chlorine
Signal Word
- Danger
Hazard Statement
- H290- May be corrosive to metals.
- H314- Causes severe skin burns and eye damage.
- H400- Very toxic to aquatic life.
- H411- Toxic to aquatic life with long lasting effects.
- EUH031- Contact with acids liberates toxic gas.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contain: Sodium hypochlorite