191.64 GBP valid until 2025-12-31
Use promo code "24372" to get your promotional price.
Use promo code "24372" to get your promotional price.
| Content And Storage |
Novex™ Sharp Pre-stained Protein Standards are provided as 2 x 250 μL (total of 50 applications of 10 μL each) of ready-to-use standard mixture. The Novex™ Sharp Pre-Stained standard loading buffer consists of 65 mM Tris pH 6.5, 30% glycerol, 2% SDS, and 2.5 mM ETDA. Store at -20°C. |
|---|---|
| Detection Method | Colorimetric |
| Number of Markers | 12 |
| Ready to Load | Yes |
| Size Range | 3.5 to 260 kDa |
| Gel Compatibility | Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels |
| Molecular Weight (g/mol) | 260, 160, 110, 80, 60, 50, 40, 30, 20, 15, 10, 3.5 kDa |
| Quantity | 2 x 250 μL |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Product Line | Novex |
| Product Type | Protein Ladder |
| Stain Type | Blue, Yellow, Pink |
| System Type | Western Blotting, SDS-PAGE |