Sodium methoxide, pure, anhydrous powder
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Overview
21.16 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 124-41-4 Weight: 0.10000kg Transport: UN number : 1431 Chem class : 4.2 Pack group : II
Quantity 100 g
Packaging Glass bottle
Disclaimers: RUO – Research Use Only
Product Code. 10687953
Quantity Price
1 £ 23.4 / 100g
In stock, ready to ship: 65
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Chemical Identifiers
CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]
Specification
Chemical Name or Material Sodium methoxide
Alkalinity total alkalinity (as NaOCH3):> 99%
CAS 124-41-4
CAS 497-19-8
Melting Point 126.0°C
Color White-Yellow
CAS 1310-73-2
Density 0.8
Packaging Glass bottle
Molecular Formula CH3NaO
Linear Formula CH3ONa
MDL Number MFCD00012179
Quantity 100 g
UN Number 1431
Beilstein 01, 280
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15, 8775
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Synonym sodium methanolate,sodium methoxide,sodium methylate,methoxysodium,methanol, sodium salt,feldalat nm,metilato sodico spanish,unii-ig663u5emc,methylate de sodium french,hsdb 755
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in methanol and ethanol, insoluble in hydrocarbons(benzene, toluene, xylene
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES C[O-].[Na+]
IUPAC Name sodium;methanolate
Molecular Weight (g/mol) 54.02
PubChem CID 10942334
Formula Weight 54.02
Percent Purity ≥97% (NaOCH3)
Grade Pure
Physical Form Powder
Alternative Products
Documentation
Technical documents
Safety and Handling
[Sodium methoxide, anhydrous powder]
[Danger]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=EYE DAM. 1, sequence=2, description=Serious eye damage/eye irritation Category 1}, {code=FLAM. SOL. 1, sequence=3, description=Flammable solid Category 1}, {code=MET. CORR. 1, sequence=4, description=Substances and mixtures corrosive to metals Category 1}, {code=SELF-HEAT. 1, sequence=5, description=Self-heating substance Category 1}, {code=SKIN CORR. 1A, sequence=6, description=Skin corrosion/irritation Category 1 A}]
[{code=H318, sequence=1, description=Causes serious eye damage.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H302, sequence=3, description=Harmful if swallowed.}, {code=H251, sequence=4, description=Self-heating: may catch fire.}, {code=H290, sequence=5, description=May be corrosive to metals.}, {code=H228, sequence=6, description=Flammable solid.}, {code=EUH014, sequence=91, description=Reacts violently with water.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P235+P410, sequence=51, description=Keep cool. Protect from sunlight.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS07, sequence=3, description=EXCLAMATION MARK}]
Product Identifier
  • Sodium methoxide, anhydrous powder
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318- Causes serious eye damage.
  • H314- Causes severe skin burns and eye damage.
  • H302- Harmful if swallowed.
  • H251- Self-heating: may catch fire.
  • H290- May be corrosive to metals.
  • H228- Flammable solid.
  • EUH014- Reacts violently with water.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235+P410- Keep cool. Protect from sunlight.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.