Potassium tert-butoxide, 98+%, pure
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Overview
13.99 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 865-47-4 Weight: 0.00500kg Transport: UN number : 3206 Chem class : 4.2 Pack group : II
Quantity 5 g
Packaging Glass Bottle
Disclaimers: RUO – Research Use Only
Product Code. 10696372
Quantity Price
1 £ 18.2 / 5g
In stock, ready to ship: 19
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Chemical Identifiers
CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.21
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]
Specification
Chemical Name or Material Potassium tert-butoxide
CAS 865-47-4
Melting Point 240.0°C to 258.0°C
Color White
Assay Percent Range 98+%
Packaging Glass Bottle
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Quantity 5 g
Fieser 01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Synonym potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in methanol, ethanol, slightly soluble in diethyl ether, benzene
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES CC(C)(C)[O-].[K+]
IUPAC Name potassium;2-methylpropan-2-olate
Molecular Weight (g/mol) 112.21
PubChem CID 23665647
Formula Weight 112.21
Percent Purity 98+%
Grade Pure
Physical Form Powder
Alternative Products
Documentation
Technical documents
Safety and Handling
[Potassium tert-butoxide]
[Danger]
[{code=FLAM. SOL. 1, sequence=1, description=Flammable solid Category 1}, {code=SELF-HEAT. 2, sequence=2, description=Self-heating substance Category 2}, {code=SKIN CORR. 1A, sequence=3, description=Skin corrosion/irritation Category 1 A}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}]
[{code=H228, sequence=1, description=Flammable solid.}, {code=H252, sequence=2, description=Self-heating in large quantities; may catch fire.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=EUH014, sequence=91, description=Reacts violently with water.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
  • Potassium tert-butoxide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Self-heating substance Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H228- Flammable solid.
  • H252- Self-heating in large quantities; may catch fire.
  • H314- Causes severe skin burns and eye damage.
  • EUH014- Reacts violently with water.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.