16.83 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Use promo code "24157" to get your promotional price.
| Capacity | 16 oz., 500 mL |
|---|---|
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Translucent |
| Description | LDPE bottle/tubulation |
| Material | LDPE |
| No. per Case | 24 |
| No. per Pack | 4 |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Shape | Cylindrical |
| Height (Metric) | 173 mm |
| Product Type | Wash Bottle |
| Height (English) | 6.81 in. |
| Closure Material | Polypropylene |
| Closure Size | 38 mm |
| Diameter (Metric) | 76.7 mm |
| Closure Type | Screw Cap |
| Through the Closure Dispensing | No |