Use promo code "24273" to get your promotional price.
Gibco™ DMEM, high glucose
DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.
17.64 GBP valid until 2025-12-31
Use promo code "24273" to get your promotional price.
Use promo code "24273" to get your promotional price.
Additional Details:
Weight:
0.59000kg
Disclaimers:
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specification
| Cell Line | HeLa, 293, Cos-7, and PC-12 |
|---|---|
| Cell Type | Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | High Glucose,Glutamine,Phenol Red |
| With Additives | High Glucose,Glutamine,Phenol Red |
| With Additives | High Glucose,Glutamine,Phenol Red |
| Without Additives | No HEPES,No Sodium Pyruvate |
| Without Additives | No HEPES,No Sodium Pyruvate |
| For Use With (Application) | Mammalian Cell Culture |
| Glucose Concentration | 4500 mg/L |
| Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
| Product Line | Gibco |
| Quantity | 500 mL |
| Shipping Condition | Room Temperature |
| Content And Storage |
Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |