Use promo code "25339" to get your promotional price.
Ammonium hydroxide, ACS reagent, 28-30% solution in water
Use promo code "25339" to get your promotional price.
| Quantity | 2.5 L |
|---|
Chemical Identifiers
| CAS | 1336-21-6 |
|---|---|
| Molecular Formula | H5NO |
| Molecular Weight (g/mol) | 35.05 |
| MDL Number | MFCD00066650 |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| SMILES | N.O |
Specification
| Chemical Name or Material | Ammonium hydroxide |
|---|---|
| Color | Colorless |
| Boiling Point | 38.0°C |
| CAS | 1336-21-6 |
| Melting Point | -57.0°C |
| CAS | 7732-18-5 |
| Molecular Formula | H5NO |
| Linear Formula | NH4OH |
| MDL Number | MFCD00066650 |
| Fieser | 05,15 |
| Density | 0.9100g/mL |
| Merck Index | 15, 491 |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| Solubility Information | Solubility in water: soluble. |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| Quantity | 2.5 L |
| SMILES | N.O |
| IUPAC Name | azanium;hydroxide |
| Molecular Weight (g/mol) | 35.05 |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| Formula Weight | 35.05 |
| Percent Purity | 28.0 to 30.0% (HCl-titration (as NH3)) |
| Grade | ACS Reagent |
| Packaging | Glass Bottle |
| Residue after Ignition | 0.002% max. |
| Specific Gravity | 0.91 |
| Vapor Pressure | 500hPa at 20°C |
Technical documents
[Ammonium hydroxide, 25-30% solution in water]
[Danger]
[{code=H314, sequence=1, description=Causes severe skin burns and eye damage.}, {code=H335, sequence=2, description=May cause respiratory irritation.}, {code=H400, sequence=3, description=Very toxic to aquatic life.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Ammonium hydroxide,Ammonia}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS09, sequence=3, description=ENVIRONMENT}]
Product Identifier
- Ammonium hydroxide, 25-30% solution in water
Signal Word
- Danger
Hazard Statement
- H314- Causes severe skin burns and eye damage.
- H335- May cause respiratory irritation.
- H400- Very toxic to aquatic life.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contain: Ammonium hydroxide,Ammonia