136.99 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Use promo code "24140" to get your promotional price.
| Barcode | No Barcode |
|---|---|
| Certified | No |
| Format | Standard |
| Detection Method | Colorimetric |
| Array | 8 x 12 |
| Packaging | 50 Cases |
| Binding Type | Slightly Hydrophilic |
| Color | Clear |
| Product Type | Microplate |
| Material | Polystyrene |
| Volume (Metric) Well | 330 μL |
| No. of Wells | 96 |
| Volume (Metric) Working | 280 μL |
| Sterility | Non-sterile |
| Surface Treatment | Immulon 2 HB |
| Well Shape | Round, Flat Bottom (F-well) |
| No. per Case | 50 |
| No. per Pack | 10 |
| Plate Blocking | None |
| Quantity | 50/Pk. |
| Target Molecule | Proteins |