Code:
66
Corning™ Reusable Polypropylene Low-Form Beakers, Graduated
Translucent polypropylene beakers printed with blue graduations.
Additional Details:
Weight:
0.23400kg
Specification
| Color | Translucent |
|---|---|
| Diameter (English) Outer | 2.63 in. |
| Diameter (Metric) Outer | 6.7 cm |
| Capacity (Metric) | 250 mL |
| Material | Polypropylene |
| For Use With (Application) | For High Temperature and Chemical Resistance |
| Height (English) | 3.7 in. |
| Height (Metric) | 9.4 cm |
| Increments | 50 mL |
| Temperature (English) Max. Operating | 140°F |
| Temperature (Metric) Max. Operating | 60°C |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Certifications/Compliance | DIN, ASTM Class A and B |
| Scale | Blue Graduated Scale |
| Style | Griffin |