Use promo code "24128" to get your promotional price.
tert-Butyl alcohol, 99%
Use promo code "24128" to get your promotional price.
| Quantity | 2500 mL |
|---|
Chemical Identifiers
| CAS | 75-65-0 |
|---|---|
| Molecular Formula | C4H10O |
| Molecular Weight (g/mol) | 74.123 |
| MDL Number | MFCD00004464 |
| InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| PubChem CID | 6386 |
| ChEBI | CHEBI:45895 |
| IUPAC Name | 2-methylpropan-2-ol |
| SMILES | CC(C)(C)O |
Specification
| Chemical Name or Material | tert-Butyl alcohol |
|---|---|
| CAS | 75-65-0 |
| Melting Point | 25°C |
| Density | 0.786 |
| Boiling Point | 83°C |
| Flash Point | 11°C (52°F) |
| Molecular Formula | C4H10O |
| Refractive Index | 1.387 |
| MDL Number | MFCD00004464 |
| Quantity | 2500 mL |
| UN Number | UN1120 |
| Beilstein | 906698 |
| Merck Index | 14,1542 |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Synonym | tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol |
| Solubility Information | Miscible with water,ethanol,ethyl ether and chloroform. |
| InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
| SMILES | CC(C)(C)O |
| IUPAC Name | 2-methylpropan-2-ol |
| Molecular Weight (g/mol) | 74.123 |
| PubChem CID | 6386 |
| ChEBI | CHEBI:45895 |
| Formula Weight | 74.12 |
| Percent Purity | 99% |
Technical documents
[tert-Butyl alcohol]
[Danger]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H332, sequence=3, description=Harmful if inhaled.}, {code=H335, sequence=4, description=May cause respiratory irritation.}, {code=H336, sequence=5, description=May cause drowsiness or dizziness.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P312, sequence=55, description=Call a POISON CENTER/doctor/if you feel unwell.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: tert-Butyl alcohol}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- tert-Butyl alcohol
Signal Word
- Danger
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H319- Causes serious eye irritation.
- H332- Harmful if inhaled.
- H335- May cause respiratory irritation.
- H336- May cause drowsiness or dizziness.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
Supplemental Information
- MIXTURE LIST- Contain: tert-Butyl alcohol