Code:
A0
DWK Life Sciences Kimble Chase™ Borosilicate Glass 3.3 Reusable Test Tubes
Borosilicate Glass 3.3 Reusable Test Tubes are available in various metric diameters and are DIN 12395 certified.
Additional Details:
Weight:
0.41400kg
Specification
| Certifications/Compliance | DIN 12395 |
|---|---|
| Design Type | Straight Neck |
| Description | Thickness: 0.8 to 1.0mm |
| Length (Metric) | 75 mm |
| Material | Borosilicate Glass |
| Disposable | Reusable |
| Quantity | 100 Pack |
| Rimmed | No |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 10 mm |
| Product Type | Reusable Test Tube |
| Bottom Shape | Round |