| Chemical Name or Material |
Potassium cyanate
|
| Melting Point |
700°C to 900°C (decomposition)
|
| CAS |
590-28-3
|
| Color |
White
|
| Physical Form |
Crystalline
|
| Quantity |
500 g
|
| Molecular Formula |
CKNO
|
| MDL Number |
MFCD00011411
|
| Merck Index |
14,7625
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Synonym |
potassium cyanate,kaliumcyanat,kocn,aero cyanate,cyanic acid, potassium salt,alicyanate,bulpur,potassiumcyanate,weedanol cyanol,caswell no. 688
|
| Solubility Information |
Soluble in water; Slightly soluble in alcohol
|
| InChI Key |
GKKCIDNWFBPDBW-UHFFFAOYSA-M
|
| SMILES |
[K+].[O-]C#N
|
| IUPAC Name |
potassium;cyanate
|
| Molecular Weight (g/mol) |
81.12
|
| PubChem CID |
11378442
|
| ChEBI |
CHEBI:38904
|
| Formula Weight |
81.12
|
| Percent Purity |
97%
|
| Odor |
Odorless
|
| Density |
2.056 g/mL
|