Use promo code "24128" to get your promotional price.
1-Methylimidazole, 99%
Use promo code "24128" to get your promotional price.
| Quantity | 100 g |
|---|
Chemical Identifiers
| CAS | 616-47-7 |
|---|---|
| Molecular Formula | C4H6N2 |
| Molecular Weight (g/mol) | 82.11 |
| MDL Number | MFCD00005292 |
| InChI Key | MCTWTZJPVLRJOU-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| PubChem CID | 1390 |
| ChEBI | CHEBI:113454 |
| SMILES | CN1C=CN=C1 |
Specification
| Chemical Name or Material | 1-Methylimidazole |
|---|---|
| CAS | 616-47-7 |
| Melting Point | -6°C |
| Density | 1.031 |
| Boiling Point | 195°C to 197°C |
| Flash Point | 92°C (197°F) |
| Molecular Formula | C4H6N2 |
| Refractive Index | 1.497 |
| MDL Number | MFCD00005292 |
| Quantity | 100 g |
| UN Number | UN3267 |
| Beilstein | 105197 |
| Sensitivity | Hygroscopic |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Synonym | 1-methyl-1h-imidazole,n-methylimidazole,1h-imidazole, 1-methyl,imidazole, 1-methyl,n-methyl imidazole,1-methyl-imidazole,unii-p4617qs63y,1-methyl-1h-imidazol,n-methylimidazol,1-methylimdazole |
| Solubility Information | Miscible with water. |
| InChI Key | MCTWTZJPVLRJOU-UHFFFAOYSA-N |
| SMILES | CN1C=CN=C1 |
| IUPAC Name | 1-methyl-1H-imidazole |
| Molecular Weight (g/mol) | 82.11 |
| PubChem CID | 1390 |
| ChEBI | CHEBI:113454 |
| Formula Weight | 82.11 |
| Percent Purity | 99% |
Technical documents
[1-Methyl-1H-imidazol]
[Danger]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H311, sequence=2, description=Toxic in contact with skin.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=4, description=Causes serious eye damage.}, {code=H361d, sequence=5, description=Suspected of damaging the unborn child.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: 1-Methyl-1H-imidazol}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
- 1-Methyl-1H-imidazol
Signal Word
- Danger
Hazard Statement
- H302- Harmful if swallowed.
- H311- Toxic in contact with skin.
- H314- Causes severe skin burns and eye damage.
- H318- Causes serious eye damage.
- H361d- Suspected of damaging the unborn child.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contain: 1-Methyl-1H-imidazol