Gibco™ TrypLE™ Express Enzyme (1X), phenol red
Animal origin-free, recombinant enzyme
Overview
308.64 GBP valid until 2025-12-31
Use promo code "24335" to get your promotional price.
Code: 50
Additional Details:
Additional Details: Weight: 0.01000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 11568856
Quantity Price
1 £ 382.0 / Pack of 20
In stock, ready to ship: 94
Add to basket
For Research Use Only. All usage must comply with product instructions.
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Specification
Chelators EDTA
Content And Storage Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 1 X
Culture Type Adherent Cell Culture
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
With Additives Phenol Red,EDTA
With Additives Phenol Red,EDTA
Osmolality Check and Record
Product Line Gibco
Quantity 20 x 100mL
Remarks Room Temperature Stable
Shipping Condition Room Temperature
Source Animal origin-free
Tested For In vitro Bioassay
Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. TrypLE™ Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts.
  • Gentle On Cells
  • urity increases specificity due to the action of single enzyme, reducing damage caused by cleavage from enzymes present in some trypsin preparations and in other protease extracts
  • Room-Temperature Stable for 24 months
  • Ready to use
  • Can be directly substituted for trypsin in existing protocols
  • Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS
Energy efficient, less hazardous, sustainable packaging
Cell Culture, Mammalian Cell Culture
Alternative Products
Documentation
Technical documents
Safety and Handling
[1-(3-Methoxyphenyl)piperazine dihydrochloride]
[Warning]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H335, sequence=3, description=May cause respiratory irritation.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P312, sequence=54, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P332+P313, sequence=55, description=If skin irritation occurs: Get medical advice/attention.}, {code=P337+P313, sequence=56, description=If eye irritation persists: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: 1-(3-Methoxyphenyl)piperazine dihydrochloride}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
  • 1-(3-Methoxyphenyl)piperazine dihydrochloride
Signal Word
  • Warning
Hazard Statement
  • H315- Causes skin irritation.
  • H319- Causes serious eye irritation.
  • H335- May cause respiratory irritation.
Precautionary Statement
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352- IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312- Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313- If skin irritation occurs: Get medical advice/attention.
  • P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
  • MIXTURE LIST- Contain: 1-(3-Methoxyphenyl)piperazine dihydrochloride