Lonza™ AccuGENE™ PBS
Ready to use solutions ideal for a wide range of molecular biology applications
Additional Details:
Weight:
1.58760kg
Specification
| Concentration | 10X |
|---|---|
| For Use With (Application) | Ideal for a wide range of molecular biology applications |
| pH | 7.4 |
| Quantity | 1000mL |
| Formulation | KH2PO4 0.017M, Na2HPO4 0.05M, NaCl 1.5M |