Elma™ EC 210 Cleaner
Ultrasonic cleaning concentrate for metal and plastic surfaces. Elma™ EC 210 Cleaner has a 3-year shelf life.
Overview
Code: NEW
Additional Details:
Additional Details: Weight: 25.50000kg
Product Code. 11741369
Quantity Price
1 £ 389.0 / 25 L
In stock, ready to ship: 8
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Specification
Form Liquid
pH 8.9
Appearance Light Beige
Boiling Point >100°C
Composition Anionic surfactants 5 to 15%, non-ionic surfactants 15 to 30%, perfumes
Concentration Ultrasonic bath 3 to 5%
Container Type Can
Content And Storage -5°C to 30°C
Volume (Metric) 25 L
  • Suitable for surfaces made of steel, nonferrous heavy metals, precious metals, light metals, nickel, chrome, zinc, and German silver (also combinations of different materials); it is also suitable for plastics that are resistant against stress cracking.
  • Check any Mg, MgZnAl alloys and plastics for compatibility before application.
  • Removes aqueous cooling lubricants, grease and oil abrasive, grinding and polishing pastes.
  • For the cleaning of polishing media containing animal greases, check the suitability of the cleaner before application.
Alternative Products
Documentation
Technical documents
Safety and Handling
[elma clean 210 (EC 210)]
[Danger]
[{code=EYE DAM. 1, sequence=1, description=Serious eye damage/eye irritation Category 1}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=AQUATIC ACUTE 1, sequence=3, description=ACUTE AQUATIC Acute 1}]
[{code=H318, sequence=1, description=Causes serious eye damage.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H400, sequence=3, description=Very toxic to aquatic life.}]
[{code=P261, sequence=51, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P312, sequence=53, description=IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.}, {code=P330, sequence=54, description=Rinse mouth.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS05, sequence=1, description=CORROSION}]
Product Identifier
  • elma clean 210 (EC 210)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H318- Causes serious eye damage.
  • H302- Harmful if swallowed.
  • H400- Very toxic to aquatic life.
Precautionary Statement
  • P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312- IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330- Rinse mouth.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.