42.35 GBP valid until 2025-07-31
Use promo code "24166" to get your promotional price.
Use promo code "24166" to get your promotional price.
| Calibration | To Deliver/Blow Out |
|---|---|
| Certifications/Compliance | Non-pyrogenic, USP XXII, USP XXII Bacterial Endotoxins Test (85); DNase/RNase Free; BSE/TSE Free |
| Material | Polystyrene |
| Sterility | Sterile |
| Graduated | Yes |
| Graduations | 0.1 mL |
| Negative Graduations | 3 mL |
| Quantity | 200/Cs., 50/Pk., 4 Packs/Cs. |
| Tolerance | ±0.2 mL |
| Packaging Type | Individual Paper/Plastic Wrap |
| Color | Orange |
| Volume (Metric) | 10 mL |
| Type | Serological Pipet |
| Disposable | Single-use |