Use promo code "24132" to get your promotional price.
Thermo Scientific™ RNase AWAY™ Surface Decontaminant
Use promo code "24132" to get your promotional price.
Specification
| pH | 11 |
|---|---|
| Composition | Sodium Hydroxide with Water |
| Container Type | Bottle |
| Volume (English) | 1.06 gal. |
| Volume (Metric) | 4 L |
Technical documents
[RNAse away]
[Warning]
[{code=H315, sequence=1, description=Causes skin irritation.}, {code=H319, sequence=2, description=Causes serious eye irritation.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P264, sequence=52, description=Wash hands thoroughly after handling.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=54, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=55, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=56, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=57, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=58, description=Immediately call a POISON CENTER/doctor/.}, {code=P363, sequence=59, description=Wash contaminated clothing before reuse.}, {code=P405, sequence=60, description=Store locked up.}, {code=P501b, sequence=61, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: sodium hydroxide}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
- RNAse away
Signal Word
- Warning
Hazard Statement
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P264- Wash hands thoroughly after handling.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
- P363- Wash contaminated clothing before reuse.
- P405- Store locked up.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
- MIXTURE LIST- Contain: sodium hydroxide