Thermo Scientific™ Legionella GVPC Selective Agar
Isolate Legionella from clinical specimens and environmental samples with a selective medium which conforms to ISO and yields optimal recovery of Legionellaceae in short incubation periods.
Overview
36.72 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Code: 64
Additional Details:
Additional Details: Weight: 0.30000kg
Product Code. 11964132
Quantity Price
1 £ 62.3 / Pack of 10
In stock, ready to ship: 7202
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Certifications/Compliance According to ISO 11731-2
Color Black, opaque
Content And Storage 2°C to 10°C
Format 90 mm Monoplate
Manufacturing Origin United Kingdom
Media Type Legionella GVPC Selective Agar
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Transmission occurs through aerosolization or aspiration of contaminated water, not person to person. Legionellosis refers to two distinct clinical syndromes, Legionnaires’ disease, which most often presents as severe pneumonia, and Pontiac fever, which is an acute, febrile, self-limiting, viral-like illness. Over forty species of Legionella have been characterized, however Legionella pneumophila remains the most prevalent etiological agent of Legionnaires’ disease. Legionella can be isolated from respiratory tract secretions of all types, sterile body fluids such as blood, and biopsy material.
This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.