Macherey-Nagel™ NucleoSpin™ TriPrep, Mini kit for RNA, DNA, and Protein Purification
Parallel isolation of RNA, DNA and, protein from an undivided sample.
Overview
344.26 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 86
Additional Details:
Additional Details: Weight: 0.15000kg Transport: UN number : 1993 Chem class : 3 Pack group : III
Product Code. 11972392
Quantity Price
1 £ 457.32 / 50 reactions
Estimated Shipment
02-06-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Specification
Filter Type Shredders
Product Type NucleoSpin TriPrep
Quantity 1 Ea.
Includes With 2mL/1.5mL Collection Tubes, NucleoSpin Filters, buffers, RNase-free rDNase
Column Type Spin Column
For Use With (Application) Rapid purification of total RNA, DNA, and protein from small and precious samples
NucleoSpin™ miRNA is suitable for the parallel isolation of small RNA, large RNA and protein.
Superior selectivity - purify your RNA fractionated by size
Isolation of small RNA only (18 to 200 bases)Isolation of small (18 to 200 bases) and large (>200 bases) RNA in two fractionsIsolation of total RNA (small and large in one fraction)
Additional isolation of total protein
Denatured protein ready-to-use for analysis in SDS-PAGE and Western blot.
Excellent RNA recovery - even without phenol/chloroform
Lysis with chaotropic saltSpin column based procedure
Convenient handling - complete equipment provided
NucleoSpin filters - efficient sample homogenisationNucleoSpin protein removal columns - highest purity of small RNArDNase - highly efficient on-column digestion of genomic DNA at RT
  • Silica membrane technology
  • Format: minispin columns
  • Sample size: <107 cultured cells, <30mg human/tissue; <50 mg plant tissue, <150μL reaction mixture
  • Fragment size, small RNA: 18 to 200 bases
  • Fragment size, large RNA: >200 bases
  • Typical yield: 10μg small RNA; 95μg large RNA from 107 HeLa cells
  • Binding capacity: 200μg
  • Elution volume, 30μL to 100μL
  • Preparation time: <45min (6 preps human/animal tissue, small and large RNA); <35min (6 preps human/animal tissue, small RNA only)
  • Small RNA (18 to 200 bases)
  • Large RNA (>200 bases)
  • Total RNA (small and large RNA in one fraction), and isolate your total protein fraction For small and large RNA and total RNA purification, and isolation of total protein fraction.
Alternative Products
Documentation
Technical documents
Safety and Handling
[NucleoSpin TriPrep]
[Danger]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H319, sequence=4, description=Causes serious eye irritation.}, {code=H334, sequence=5, description=May cause allergy or asthma symptoms or breathing difficulties if inhaled.}, {code=H412, sequence=6, description=Harmful to aquatic life with long lasting effects.}]
[{code=P261, sequence=51, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P342+P311, sequence=52, description=If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .}]
[{code=MIXTURE LIST, sequence=1, description=Contain: zinc chloride; guanidinium thiocyanate,ethanol (denatured with 1%IPA/1%MEK, acc.2016/1867/EU); rDNa}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
  • NucleoSpin TriPrep
Signal Word
  • Danger
Hazard Statement
  • H226- Flammable liquid and vapour.
  • H302- Harmful if swallowed.
  • H315- Causes skin irritation.
  • H319- Causes serious eye irritation.
  • H334- May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412- Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311- If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental Information
  • MIXTURE LIST- Contain: zinc chloride; guanidinium thiocyanate,ethanol (denatured with 1%IPA/1%MEK, acc.2016/1867/EU); rDNa