Use promo code "25339" to get your promotional price.
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™
Use promo code "25339" to get your promotional price.
| Quantity | 2.5 L |
|---|---|
| Packaging | AcroSeal™ Glass Bottle |
Chemical Identifiers
| CAS | 96-47-9 |
|---|---|
| Molecular Formula | C5H10O |
| Molecular Weight (g/mol) | 86.13 |
| MDL Number | MFCD00005367 |
| InChI Key | JWUJQDFVADABEY-UHFFFAOYSA-N |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| PubChem CID | 7301 |
| IUPAC Name | 2-methyloxolane |
| SMILES | CC1CCCO1 |
Specification
| Chemical Name or Material | 2-Methyltetrahydrofuran |
|---|---|
| Melting Point | -136.0°C |
| Name Note | Stabilized |
| CAS | 96-47-9 |
| CAS Min % | 99.0 |
| Color | Colorless |
| CAS Max % | 100.0 |
| Density | 0.8600g/mL |
| Quantity | 2.5 L |
| Infrared Spectrum | Authentic |
| Packaging | AcroSeal™ Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.4046 to 1.4066 |
| Assay Percent Range | 99% min. (GC) |
| Specific Gravity | 0.86 |
| Molecular Formula | C5H10O |
| MDL Number | MFCD00005367 |
| Viscosity | 4 mPa.s (25°C) |
| UN Number | 2536 |
| Merck Index | 15,6198 |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Synonym | 2-methyltetrahydrofuran,methyltetrahydrofuran,tetrahydrosylvan,tetrahydro-2-methylfuran,furan, tetrahydro-2-methyl,furan, 2-methyl-tetrahydro,2-methyl tetrahydrofuran,2-methyl-tetrahydrofuran,2-methyl-tetrahydro-furan,methyl tetrahydrofuran |
| Boiling Point | 78.0°C to 80.0°C |
| Solubility Information | Solubility in water: 150g/l (25°C) |
| InChI Key | JWUJQDFVADABEY-UHFFFAOYSA-N |
| SMILES | CC1CCCO1 |
| IUPAC Name | 2-methyloxolane |
| Molecular Weight (g/mol) | 86.13 |
| PubChem CID | 7301 |
| Formula Weight | 86.13 |
| Percent Purity | 99+% |
| Grade | Extra Dry |
Technical documents
[2-Methyltetrahydrofuran]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=SKIN IRRIT. 2, sequence=3, description=Skin corrosion/irritation Category 2}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H318, sequence=4, description=Causes serious eye damage.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P305+P351+P338, sequence=52, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=53, description=IF exposed or concerned: Get medical advice/attention.}, {code=P210, sequence=54, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS07, sequence=3, description=EXCLAMATION MARK}]
Product Identifier
- 2-Methyltetrahydrofuran
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H302- Harmful if swallowed.
- H315- Causes skin irritation.
- H318- Causes serious eye damage.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.