Fisherbrand™ 0.2mL PCR Tubes
With integral snap shut cap
Overview
32.91 GBP valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Code: LC
Additional Details:
Additional Details: Weight: 0.19800kg
Product Code. 12134102
Quantity Price
1 £ 67.38 / Pack of 1000
In stock, ready to ship: 106
In stock, ready to ship: 45
Specification
Certifications/Compliance RNase, Dnase and human DNA free
Color Natural
Volume (Metric) 0.2 mL
Quantity 1000 Case
For Use With (Equipment) 0.2mL thermal cycler blocks for PCR and sample storage applications
Material Polypropylene
Closure Type Flat Cap
Format Tube
Product Type Thin Wall PCR Tube
  • Suitable for 0.2mL thermal cycler blocks
  • 0.25mL maximum volume when closed
  • Thin-wall design
  • Cleanroom produced
  • Virgin polypropylene
Alternative Products