Eppendorf™ Centrifuge 5430/5430 R
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
7146.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Code:
99
Additional Details:
Weight:
57.00000kg
Specification
| Capacity | 30 x 1.5/2 mL |
|---|---|
| Max. RCF | 30,130 x g |
| Refrigerated | Yes |
| Display | Large, brightly lit LCD |
| Height (English) Exterior | 11.7 in. |
| Height (Metric) Exterior | 29 cm |
| Noise Level | <54 dB(A) with Rotor F-45-30-11 |
| Safety Features | Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition |
| Timer | 30 sec. to 99:59 hrs., with continuous run function |
| Width (English) Exterior | 15 in. |
| Width (Metric) Exterior | 38 cm |
| Wattage | 1.050 W |
| Certifications/Compliance | UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers |
| Type | Centrifuge |
| Includes | With Rotor FA-45-30-11 incl. rotor lid |
| Electrical Requirements | 230 V, 50/60 Hz (GB) |
| Controller Type | Rotary Knob |
| Depth (Metric) | 640 mm |
| Maximum Speed Range | 100 to 17,500 rpm |
| Net Weight | 56 kg |
| Run Time | 30 sec. to 9 hr. 59 min. |
| Standards | EN ISO 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-1, EN ISO 18113-3, EN ISO 15223-1, EN 61010-1, EN 61010-2-020, IEC 61010-1, IEC 61010-2-020, EN 55011, EN 61326-1, EN 50581 (refer to manual for more details) |
| Temperature Range | -11°C to +40°C |
| Frequency | 50/60 Hz |
| Voltage | 230 V |
| Model | Centrifuge 5430 R (EU-IVD) |