1296.39 GBP valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Use promo code "24148" to get your promotional price.
| Description | MicroClick 24 x 2 Fixed Angle Microtube Rotor |
|---|---|
| Type | Fixed Angle Rotor |
| Capacity | 24 x 2 mL |
| Max. RCF | 24,372 x g |
| Max. Speed | 16,000 rpm |
| Radius (Metric) | 85 mm |
| K Factor | 505 |
| Max. Tube Length | 45 mm |
| Max. Tube Diameter | 11 mm |
| For Use With (Equipment) | Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges |
| Angle | 45° |
| Biocontainment | Yes |