Thermo Scientific™ Sorvall™ ST 8 Small Benchtop Centrifuge
Meet space, clinical and research protocal needs with the compact and innovative Thermo Scientific™ Sorvall™ ST 8 Small Benchtop Centrifuge. Rotors and buckets are sold separately.
5805.57 GBP valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Use promo code "24148" to get your promotional price.
Code:
NEW
Additional Details:
Weight:
71.00000kg
Disclaimers:
NOTE: Rotors and buckets must be purchased separately.
Specification
| Capacity | 4 x 145 mL (TX-150), 6 x 50 mL (HIGHConic III) |
|---|---|
| Max. Speed | 4,500 rpm (TX-150 rotor), 17,850 rpm (MicroClick 24 x 2 rotor) |
| Max. RCF | 3,260 x g (TX-150 Rotor); 30,279 x g (MicroClick 24 x 2 Rotor) |
| Refrigerated | Yes |
| Height (English) Exterior | 12.6 in. |
| Height (Metric) Exterior | 32 cm |
| Length (English) Exterior | 26.4 in. |
| Length (Metric) Exterior | 67 cm |
| Noise Level | <52 dB(A) (with MicroClick 24 x 2 rotor); <56 dB(A) with all rotors |
| Safety Features | Auto-Lock rotor exchange system, SMARTSpin imbalance detection system, certified ClickSeal biocontainment lids |
| Width (English) Exterior | 18.1 in. |
| Width (Metric) Exterior | 46 cm |
| Wattage | 750 w |
| Certifications/Compliance | IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1; UL Listed / CE marked /IVD compliant / Biocontainment certified by the Public Health England, Porton Down, UK |
| Includes | ST 8 Centrifuge. (Rotors and buckets are sold separately.) |
| Description | Sorvall ST 8R Small Benchtop Centrifuge |
| Electrical Requirements | 230 V 50/60 Hz |
| Controller Type | Microprocessor |
| Rotor Included | No |
| Dimensions (HWD) | 12.6 x 18.1 x 26.4 in. (32 x 46 x 67 cm) |
| Drive System | Direct Brushless Induction |
| Net Weight | 156 lb. (71 kg) |
| Program Storage | 3 direct program buttons, plus 96 additional programs accessible via folder, all with alphanumeric program naming |
| Run Time | 99 hr. 59 min., continuous |
| Temperature Range | -10°C to +40°C |
| Product Line | Sorvall |
| Height (English) Exterior Lid Open | 27.5 in. |
| Profile (Acceleration/Braking) | 2 (standard and soft) |
| Frequency | 50/60 Hz |
| Height (Metric) Exterior Lid Open | 70 cm |
| Rotor | TX-150 |
| Voltage | 230 V |