Use promo code "25339" to get your promotional price.
Thermo Scientific Chemicals Guanidine hydrochloride, 99+%
Use promo code "25339" to get your promotional price.
| Quantity | 1 kg |
|---|
Chemical Identifiers
| CAS | 50-01-1 |
|---|---|
| Molecular Formula | CH6ClN3 |
| Molecular Weight (g/mol) | 95.53 |
| MDL Number | MFCD00013026 |
| InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| PubChem CID | 5742 |
| ChEBI | CHEBI:32735 |
| IUPAC Name | guanidine;hydrochloride |
| SMILES | C(=N)(N)N.Cl |
Specification
| Chemical Name or Material | Guanidine Hydrochloride |
|---|---|
| CAS | 50-01-1 |
| Melting Point | 185°C to 188°C |
| Color | White |
| Density | 1.344 g/mL |
| Molecular Formula | CH6ClN3 |
| MDL Number | MFCD00013026 |
| Quantity | 1 kg |
| Beilstein | 3591990 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,4562 |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Synonym | guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride |
| Solubility Information | Freely soluble in water;Soluble in water and ethanol. |
| InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
| SMILES | C(=N)(N)N.Cl |
| IUPAC Name | guanidine;hydrochloride |
| Molecular Weight (g/mol) | 95.53 |
| PubChem CID | 5742 |
| ChEBI | CHEBI:32735 |
| Formula Weight | 95.53 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
Technical documents
[Guanidine hydrochloride]
[Warning]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H319, sequence=3, description=Causes serious eye irritation.}, {code=H332, sequence=4, description=Harmful if inhaled.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=52, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P312, sequence=55, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P337+P313, sequence=56, description=If eye irritation persists: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: Guanidine, hydrochloride}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
- Guanidine hydrochloride
Signal Word
- Warning
Hazard Statement
- H302- Harmful if swallowed.
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H332- Harmful if inhaled.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contain: Guanidine, hydrochloride