4-(Dimethylamino)pyridine, 99%, prilled
CAS: 1122-58-3 | C7H10N2 | 122.171 g/mol
Overview
114.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 1122-58-3 Weight: 0.10000kg Transport: UN number : 2811 Chem class : 6.1 Pack group : II
Quantity 100 g
Disclaimers: RUO – Research Use Only
Product Code. 15448167
Quantity Price
1 £ 114.0 / 100g
Estimated Shipment
20-07-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 1122-58-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006418
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
PubChem CID 14284
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)C1=CC=NC=C1
Specification
Chemical Name or Material 4-(Dimethylamino)pyridine
CAS 1122-58-3
Melting Point 110°C to 113°C
Boiling Point 162°C (50 mmHg)
Flash Point 124°C (255°F)
Molecular Formula C7H10N2
MDL Number MFCD00006418
Quantity 100 g
UN Number UN2928
Beilstein 110354
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Synonym 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine
Solubility Information Soluble in methanol,benzene,ethyl acetate,chloroform,methylene chloride,acetone,and acetic acid. Slightly soluble in ether,diisopropyl ether,cyclohexane,hexane,and water.
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
SMILES CN(C)C1=CC=NC=C1
IUPAC Name N,N-dimethylpyridin-4-amine
Molecular Weight (g/mol) 122.171
PubChem CID 14284
Formula Weight 122.17
Percent Purity 99%
Physical Form Prills
Applications
4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.
Solubility
Soluble in methanol, benzene, ethyl acetate, chloroform, methylene chloride, acetone, and acetic acid. Slightly soluble in ether, diisopropyl ether, cyclohexane, hexane, and water.
Notes
Incompatible with oxidizing agents and acids.