134.55 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
| ANSI Abrasion Rating | ANSI 6 |
|---|---|
| Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
| Construction | Knit |
| Coating Material | Polyurethane |
| Color | White,Black |
| Color | White,Black |
| Cut Resistance Rating | ANSI A4 |
| Hand | Specific |
| Latex Free | Yes |
| Material | Fiberglass,HPPE,Nylon,Spandex,Polyethylene |
| Material | Fiberglass,HPPE,Nylon,Spandex,Polyethylene |
| Material | Fiberglass,HPPE,Nylon,Spandex,Polyethylene |
| Material | Fiberglass,HPPE,Nylon,Spandex,Polyethylene |
| Material | Fiberglass,HPPE,Nylon,Spandex,Polyethylene |
| Product Type | Cut-resistant Gloves |
| Size | 7 |
| EN388 Standard | EN 388:4X43C, EN 388:4543 |
| For Use With (Application) | Assembly of small parts and components, cutting of small dry or lightly oiled parts and maintenance, general maintenance, metal fabrication |
| Gender | Unisex |
| Glove Cuff Style | Knit |
| Grip Design | Rough |
| Liner Material | Fiberglass, HPPE, Nylon, Spandex |
| Lining | Lined |
| Model | 11-735 7 163843 |
| Packaging Type | Case |
| Puncture Rating | ANSI/ISEA 105-16 Level 3 |
| Series | HyFlex™ 11-735 |
| Thumb Style | Wing |
| Length (English) | 8.8 in. |
| Length (Metric) | 22.4 cm |