Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 with Certificate
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Additional Details:
Weight:
5.00000kg
Specification
| Classification | E2 |
|---|---|
| Material | Stainless Steel |
| Weight (Metric) | 5 kg |
| Mass | 5 kg |
| Shape | Cylindrical |