HUBER KISS K6 Refrigerated Heating Bath
Cooling bath thermostats with insulated cooling baths are an economic solution for thermoregulation of objects directly in the thermostat bath. The units are suitable for a typical heating and cooling tasks such as sample temperature control, materials testing, quality control, analyses, etc.
4210.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Additional Details:
Weight:
25.00000kg
Specification
| Controller Type | KISS |
|---|---|
| Capacity (Metric) | 4.5 L |
| Cooling Capacity | 20°C to -20°C |
| Display | OLED |
| Heating Capacity | 1.6 - 2.1 kW |
| Max. Pressure | 14 L/min; 0.25 bar |
| Model | KISS K6 |
| Pump Capacity | M16x1 AG male |
| Safety Class | 3/FL (DIN 12876) |
| Voltage | 208/240 V |
| Frequency | 50/60 Hz |