Code:
NEW
Ethylene glycol, Reagent Grade, ≥99%, Solstice
Reagent Grade,≥99%
Additional Details:
CAS Number:
107-21-1
Weight:
22.26000kg
| Quantity | 20 L |
|---|---|
| Packaging | Stainless Steel Can |
Chemical Identifiers
| CAS | 107-21-1 |
|---|---|
| Molecular Formula | C2H6O2 |
| Molecular Weight (g/mol) | 62.068 |
| MDL Number | MFCD00002885 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| PubChem CID | 174 |
| ChEBI | CHEBI:30742 |
| IUPAC Name | ethane-1,2-diol |
| SMILES | C(CO)O |
Specification
| Chemical Name or Material | Ethylene glycol |
|---|---|
| Melting Point | -13°C (literature) |
| Name Note | Reagent Grade, ≥99% |
| CAS | 107-21-1 |
| CAS Max % | 100.0000% |
| Density | 1.113 g/mL at 25 °C(lit.) |
| Quantity | 20 L |
| Flash Point | 111°C |
| Packaging | Stainless Steel Can |
| Molecular Formula | C2H6O2 |
| Linear Formula | HOCH2CH2OH |
| MDL Number | MFCD00002885 |
| Beilstein | 505945 |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Synonym | ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol |
| Boiling Point | 197°C |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| SMILES | C(CO)O |
| IUPAC Name | ethane-1,2-diol |
| Molecular Weight (g/mol) | 62.068 |
| PubChem CID | 174 |
| ChEBI | CHEBI:30742 |
| Formula Weight | 62.07 |
| Percent Purity | >99% |
| Grade | Reagent |