19.66 GBP valid until 2025-07-22
Use promo code "24098" to get your promotional price.
Use promo code "24098" to get your promotional price.
| Certifications/Compliance | NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600 |
|---|---|
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, EN 455 1-4 |
| Accelerator Free | No |
| Applications | Healthcare,Laboratory |
| Applications | Healthcare,Laboratory |
| AQL | 0.65 |
| Color | Black |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Finger Thickness (English) | 6.3 mil |
| Finger Thickness (Metric) | 0.16 mm |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 4.7 mil |
| Palm Thickness (Metric) | 0.12 mm |
| Powdered | Powder Free |
| Product Type | Nitrile Examination Glove |
| Size | Large |
| Sterility | Non-sterile |
| Thickness (English) | 6.3 mil |
| Thickness (Metric) | 0.16 mm |
| Model | 93862090 |
| Series | MICROFLEX™ MidKnight™ XTRA 93-862 |
| Length (English) | 12 in. |
| Length (Metric) | 30 cm |