Fisherbrand™ White Polypropylene Caps
White polypropylene screw caps provides clear surface for writing on cap
Additional Details:
Weight:
0.94500kg
Specification
| Color | White |
|---|---|
| Material | Polypropylene |
| Quantity | 370/Cs. |
| Liner Material | Polyethylene Foam |
| Neck Finish | 89-400 |
| Product Type | Lined Cap |