Labcon ZAP™ 20 μL Aerosol Filter Pipette Tips
Fits Most Popular 20 μL Pipettors
Additional Details:
Weight:
3.62870kg
Specification
Volume (Metric) | 20 μL |
---|---|
Filtered | Filtered |
Sterility | Sterile |
Color | Natural |
Autoclavable | Not Autoclavable |
Length (English) | 1.95 in. |
Length (Metric) | 49.7 mm |
Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity |
Clean Claims | Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) |
For Use With (Application) | Liquid Handling |
Sterilization Method | E-beam |
Format | Bagged |
Tip Style | Beveled |
Packaging | 1 Tip/Bag |
Shipping Condition | RT |
Disposable | Single-use |
For Use With (Equipment) | Universal Fit |
Graduated | No |
Product Line | ZAP |
Material | Polypropylene Resin |