Invitrogen™ xMAP Sheath Fluid Plus
xMAP Sheath Fluid Plus is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments.
Overview
67.21 GBP valid until 2025-08-31
Use promo code "24128" to get your promotional price.
Additional Details:
Additional Details: Weight: 20.00000kg
Product Code. 17183935
Quantity Price
1 £ 73.0 / 20 L
In stock, ready to ship: 12
Add to basket
Description and Specification
Specification
Quantity 20 L
Packaging Container
xMAP Sheath Fluid Plus is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments. It is needed to operate all Luminex 200 and FLEXMAP 3D systems.
This reformulated sheath product replaces the original xMAP Sheath Fluid (Cat. No. 4050015). The new sheath formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
For Research Use Only. Not for use in diagnostic procedures. Not intended for human or animal therapeutic or diagnostic use.