VEE GEE Thermometer, Encapsulated in FEP with Armor
The armored encapsulated glass thermometer is protected in the event of breakage, the FEP coating contains the broken glass and liquid fill, preventing injury or contamination. The armor protects the thermometer from rough handling and shock during use and encapsulation protects you.
Additional Details:
CAS Number:
00-00-0
Weight:
0.35000kg
Specification
| Bottle Fill | Safety Blue |
|---|---|
| For Use With (Application) | Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications |
| Immersion Type | Partial Immersion |
| Length (Metric) | 30.48 cm |
| Material | Glass |
| Probe Type | Immersion/General Purpose (Tapered) |
| Thermometer Fill | Safety Blue |
| Weight (Metric) | 11 g |
| Product Type | Serialized Glass Thermometer |
| Description | Nickel-plated brass armor |
| Max. Temperature (English) | 230°F |
| Max. Temperature (Metric) | 110°C |
| Min. Temperature (English) | -4°F |
| Min. Temperature (Metric) | -20°C |
| Temperature Range (English) | -4 to +230°F |
| Temperature Range (Metric) | -20 to 110°C |
| Accuracy | ±1 subdivision |
| Subdivision | 1°C (1.8°F) |
| Immersion Length | To immersion line |
| Width (Metric) | 1.65 cm |