N-tert-Butoxycarbonyl-1,6-hexanediamine, 95%
CAS: 51857-17-1 | C11H24N2O2 | 216.33 g/mol
Overview
220.38 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 51857-17-1 Weight: 0.00500kg Transport: UN number : 2735 Chem class : 8 Pack group : III
Quantity 5 g
Packaging Plastic Bottle
Disclaimers: RUO – Research Use Only
Product Code. 10560404
Quantity Price
1 £ 220.0 / 5g
Estimated Shipment
21-04-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 51857-17-1
Molecular Formula C11H24N2O2
Molecular Weight (g/mol) 216.33
MDL Number MFCD00671489
InChI Key RVZPDKXEHIRFPM-UHFFFAOYSA-N
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
PubChem CID 2733170
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCCN
Specification
Chemical Name or Material N-tert-Butoxycarbonyl-1, 6-hexanediamine
CAS 51857-17-1
Boiling Point 106°C to 110°C (0.3mmHg)
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Molecular Formula C11H24N2O2
Packaging Plastic Bottle
Refractive Index 1.4590 to 1.461
MDL Number MFCD00671489
UN Number 2735
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Synonym n-boc-1,6-diaminohexane,n-boc-1,6-hexanediamine,tert-butyl n-6-aminohexyl carbamate,tert-butyl 6-aminohexyl carbamate,n-tert-butoxycarbonyl-1,6-diaminohexane,n-tert-butoxycarbonyl-1,6-hexanediamine,tert-butyl 6-aminohexylcarbamate,boc-1,6-diaminohexane hydrochloride,6-tert-butoxycarbonylamino hexylamine,n-6-aminohexyl carbamic acid tert-butyl ester
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,ch2cl2
InChI Key RVZPDKXEHIRFPM-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NCCCCCCN
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate
Molecular Weight (g/mol) 216.33
Quantity 5 g
Flash Point 125°C
PubChem CID 2733170
Formula Weight 216.33
Percent Purity 95%
Physical Form Viscous Liquid