37.28 GBP valid until 2025-07-22
Use promo code "24098" to get your promotional price.
Use promo code "24098" to get your promotional price.
| Certifications/Compliance | ISO 3,ISO 4,ISO 5,ISO 6,ISO 7 |
|---|---|
| Certifications/Compliance | ISO 3,ISO 4,ISO 5,ISO 6,ISO 7 |
| Certifications/Compliance | ISO 3,ISO 4,ISO 5,ISO 6,ISO 7 |
| Certifications/Compliance | ISO 3,ISO 4,ISO 5,ISO 6,ISO 7 |
| Certifications/Compliance | ISO 3,ISO 4,ISO 5,ISO 6,ISO 7 |
| Certifications/Compliance (Europe) | GMP Grade A/B/C/D |
| Chemotherapeutic Rated | ASTM D6978 |
| Chlorinated | Yes |
| Cleanroom Class | Class 10 (ISO 4) |
| Accelerator Free | Yes |
| AQL | 1 |
| Color | White |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Finger Thickness (English) | 6 mil |
| Finger Thickness (Metric) | 0.16 mm |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 5 mil |
| Palm Thickness (Metric) | 0.13 mm |
| Powdered | Powder Free |
| Product Type | Controlled Environment Gloves |
| Size | Small |
| Sterility | Non-sterile |
| Thickness (English) | 6 mil |
| Thickness (Metric) | 0.16 mm |
| Manufacturing/QMS Audit Standards | ISO 9001:2015, EN13485:2016 |
| Length (English) | 12 in. |
| Length (Metric) | 30.5 cm |