1,3-Dimethyl-2-imidazolidinone, 98%
CAS: 80-73-9 | C5H10N2O | 114.15 g/mol
£25.4 - £152.0
Chemical Identifiers
CAS 80-73-9
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD00003188
InChI Key CYSGHNMQYZDMIA-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
PubChem CID 6661
IUPAC Name 1,3-dimethylimidazolidin-2-one
SMILES CN1CCN(C)C1=O
View More Specs
Disclaimers: RUO – Research Use Only
Specification
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone
CAS 80-73-9
Melting Point 8°C
Boiling Point 224°C to 226°C
Flash Point 93°C (199°F)
Molecular Formula C5H10N2O
MDL Number MFCD00003188
Quantity 25 g
UN Number UN2810
Beilstein 108808
Merck Index 14,3249
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
Synonym 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
InChI Key CYSGHNMQYZDMIA-UHFFFAOYSA-N
SMILES CN1CCN(C)C1=O
IUPAC Name 1,3-dimethylimidazolidin-2-one
Molecular Weight (g/mol) 114.15
PubChem CID 6661
Formula Weight 114.15
Percent Purity 98%
Applications
1,3-Dimethyl-2-imidazolidinone is used as a high-boiling polar aprotic solvent. It is used as a substitute solvent for toxic hexamethylphosphoramide. It finds application in detergents, dyestuffs, electronic materials and in the manufacture of polymers. It is involved in the preparation of 1,2-bis(trimethylsilyl)benzene. Further, it is utilized as a solvent during alfa-regioselective prenylation of imines. In addition to this, it constitutes the mobile phase during size-exclusion chromatographic analysis of cellulose.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides and phosphorus halides.
Product Code Brand Quantity Price ??product.searchResults.label.quantityAvail??
Quantity
11488596
Safety data sheet (SDS)
N/A
A16001.14
25 g £25.4
25g
Add to basket
11498596
Safety data sheet (SDS)
N/A
A16001.22
100 g £47.0
100g
Add to basket
11408606
Safety data sheet (SDS)
N/A
A16001.36
500 g £152.0
500g
Add to basket
Product Identifier
  • 1,3-Dimethyl-2-imidazolidinone
Signal Word
  • Danger
Hazard Statement
  • H302- Harmful if swallowed.
  • H318- Causes serious eye damage.
  • H361- Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201- Obtain special instructions before use.
  • P202- Do not handle until all safety precautions have been read and understood.
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264- Wash face, hands, and anyexposed skin thoroughly after handling.
  • P270- Do not eat, drink or smoke when using this product.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.
  • P330- Rinse mouth.
  • P370+P378- In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
  • P403+P235- Store in a well-ventilated place. Keep cool.
  • P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
  • MIXTURE LIST- Contain: 1,3-Dimethylimidazolidin-2-one